Anti-PURH Rabbit Polyclonal Antibody

Supplier: Proteintech
10726-1-AP-150UL
10083-258EA 459.7 USD
10083-258
Anti-PURH Rabbit Polyclonal Antibody
Antibodies
Western Blot: HeLa Cells, 1:200-1:1000; IF: HeLa Cells, 1:10-1:100

Type: Primary
Antigen: PURH
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR