Anti-CCNH Rabbit Polyclonal Antibody

Supplier: Proteintech
10718-1-AP-150UL
10085-480EA 459.7 USD
10085-480
Anti-CCNH Rabbit Polyclonal Antibody
Antibodies
Western Blot:A375 Cells, 1:500-1:5000; IHC: Human ovary tumor Tissue, 1:20-1:200; IF:SH-SY5Y Cells, 1:20-1:200; FC:SH-SY5Y Cells, N/A

Type: Primary
Antigen: CCNH
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR