Anti-SIX1 Rabbit Polyclonal Antibody

Supplier: Proteintech
10709-1-AP-150UL
10094-418EA 459.7 USD
10094-418
Anti-SIX1 Rabbit Polyclonal Antibody
Antibodies
SIX1 belongs to SIX family that contains a divergent DNA-binding homeodomain and an upstream SIX domain, which may participate both in determining DNA-binding specificity and in mediating protein-protein interactions. It may be involved in limb tendon and ligament development, and specifically expressed in skeletal muscle. SIX1 plays a essential role in the development of numerous organs and shows littlt to no expression in most non-neoplastic adult tissue, yet is overexpression in a number of meoplasms where it increases cell proliferation and survival.

Western Blot:A2780 Cells, 1:200-1:2000; IF:epithelial tumor Cells, 1:20-1:200; IP: HEK-293 Cells, 1:500-1:5000

Type: Primary
Antigen: SIX1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR