Anti-HSF2 Rabbit Polyclonal Antibody

Supplier: Proteintech
10706-1-AP-150UL
10088-492EA 459.7 USD
10088-492
Anti-HSF2 Rabbit Polyclonal Antibody
Antibodies
IHC: Human prostate cancer, 1:20-1:200

Type: Primary
Antigen: HSF2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR