Anti-RGS2 Rabbit Polyclonal Antibody

10678-1-AP
10093-624EA 356.38 USD
10093-624
Anti-RGS2 Rabbit Polyclonal Antibody
Western Blot: Human Brain Tissue, 1:200-1:2000; IHC: Human pancreas cancer, 1:20-1:200

Type: Primary
Antigen: RGS2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR