Anti-OSTF1 Rabbit Polyclonal Antibody

Supplier: Proteintech
10671-1-AP-150UL
10091-628EA 459.7 USD
10091-628
Anti-OSTF1 Rabbit Polyclonal Antibody
Antibodies
OSTF1 (Osteoclast-stimulating factor-1) is an intracellular protein produced by osteoclasts that indirectly induces osteoclast formation and bone resorption. It induces bone resorption, acting probably through a signaling cascade which results in the secretion of factor(s) enhancing osteoclast formation and activity

IHC: Human breast cancer, 1:20-1:200

Type: Primary
Antigen: OSTF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR