HSD17B10 (3-hydroxyacyl-CoA dehydrogenase type-2) is a multifunctional mitochondrial enzyme that acts on a wide spectrum of substrates, including neuroactive steroids, alcohols, leucine, and fatty acids, with a preference for short-chain methyl-branched acyl-CoAs.It has 2 isoforms produced by alternative splicing.Defects in HSD17B10 are the cause of 2-methyl-3-hydroxybutyryl-CoA dehydrogenase deficiency (MHBD deficiency) and mental retardation syndromic X-linked type 10 (MRXS10).
Western Blot: HeLa Cells, 1:500-1:5000; IHC: Human breast cancer Tissue, 1:20-1:200; FC: MCF-7 Cells, N/A
Type: Primary
Antigen: ERAB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat