Anti-TOR3A Rabbit Polyclonal Antibody

Supplier: Proteintech
10647-1-AP-150UL
10096-208EA 459.7 USD
10096-208
Anti-TOR3A Rabbit Polyclonal Antibody
Antibodies
Western Blot: HEK-293 Cells, 1:200-1:1000

Type: Primary
Antigen: TOR3A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR