Anti-RHOC Rabbit Polyclonal Antibody

Supplier: Proteintech
10632-1-AP-150UL
10093-652EA 459.7 USD
10093-652
Anti-RHOC Rabbit Polyclonal Antibody
Antibodies
Ras homolog family member C (RHOC) encodes a Rho-related GTP-binding protein, which belongs to small GTPase superfamily and also Rho family. RHOC. Regulates a signal transduction pathway linking plasma membrane receptors to the assembly of focal adhesions and actin stress fibers. Expression of RHOC contributes to metastatic phenotype of melanoma cells and is associated with multiple cancers including gastric carcinomas, ovarian carcinomas and breast cancer as well.

Western Blot: HeLa Cells, 1:200-1:2000

Type: Primary
Antigen: RHOC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR