Anti-FASN Rabbit Polyclonal Antibody

Supplier: Proteintech
10624-2-AP-150UL
10086-878EA 599.76 USD
10086-878
Anti-FASN Rabbit Polyclonal Antibody
Antibodies
Western Blot:rat Liver Tissue, 1:200-1:1000; IF: Hela Cells, 1:10-1:100; IP: Mouse Liver Tissue, 1:200-1:2000; FC: HeLa Cells, N/A

Type: Primary
Antigen: FASN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR