Anti-FUBP3 Rabbit Polyclonal Antibody

Supplier: Proteintech
10623-1-AP-150UL
10087-174EA 459.7 USD
10087-174
Anti-FUBP3 Rabbit Polyclonal Antibody
Antibodies
IHC: Human prostate cancer, 1:20-1:200

Type: Primary
Antigen: FUBP3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR