Anti-AP1M2 Rabbit Polyclonal Antibody

Supplier: Proteintech
10618-1-AP-150UL
10082-964EA 459.7 USD
10082-964
Anti-AP1M2 Rabbit Polyclonal Antibody
Antibodies
AP1M2 is a subunit of the adaptor complex AP-1. It belongs to the adaptor complexes medium subunit family. Adaptor protein (AP) complexes are cytosolic heterotetramers that mediate the sorting of membrane proteins in the secretory and endocytic pathways. AP-1 is found at the cytoplasmic face of coated vesicles located at the Golgi complex, where it mediates both the recruitment of clathrin to the membrane and the recognition of sorting signals within the cytosolic tails of transmembrane receptors.

Western Blot: Mouse Colon Tissue, 1:200-1:2000; IHC: Human pancreas cancer Tissue, 1:10-1:100

Type: Primary
Antigen: AP1M2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR