Anti-CKS1B Rabbit Polyclonal Antibody

10610-1-AP
10084-814EA 340.25 USD
10084-814
Anti-CKS1B Rabbit Polyclonal Antibody
Type: Primary
Antigen: CKS1B
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR