Anti-PLCD4 Rabbit Polyclonal Antibody

Supplier: Proteintech
10589-2-AP
10092-436EA 360.13 USD
10092-436
Anti-PLCD4 Rabbit Polyclonal Antibody
Antibodies
IHC: Human pancreas cancer, 1:20-1:200

Type: Primary
Antigen: PLCD4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR