Anti-TRAPA Rabbit Polyclonal Antibody

Supplier: Proteintech
10583-1-AP-150UL
10096-280EA 459.7 USD
10096-280
Anti-TRAPA Rabbit Polyclonal Antibody
Antibodies
TRAPA (translocon-associated protein subunit alpha), also known as SSR1 (signal sequence receptor subunit alpha), is a 34-kDa single-spanning membrane glycoprotein of the endoplasmic reticulum (ER) . It is a part of the tetrameric TRAP complex (TRAP-alpha, TRAP-beta, TRAP-delta and TRAP-gamma) which functions in regulating the retention of ER resident proteins . The TRAP complex is also involved in endoplasmic reticulum-associated degradation (ERAD) .

Western Blot: Human Liver Tissue, 1:200-1:1000; IHC: Human ovary tumor Tissue, 1:20-1:200

Type: Primary
Antigen: TRAPA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR