Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-HNRNPUL1 Rabbit Polyclonal Antibody

Supplier: Proteintech
HNRNPUL1, also named as E1BAP5, HNRPUL1 and E1B-AP5, acts as a basic transcriptional regulator. It represses basic transcription driven by several virus and cellular promoters. HNRNPUL1 plays also a role in mRNA processing and transport. It is a cellular protein that interacts with the adenovirus protein E1B-55 and implicated in mRNA export. HNRNPUL1 is an hnRNP-like protein that likely mediates the interaction of NXF1 with mRNAs.. It may modulate PAN expression in complex with KSHV ORF57. HNRNPUL1 specifically bound to the Rae1–Nup98 complex independently of M protein, and this interaction was found to be RNA dependent, as it was greatly diminished in the presence of RNase. HNRNPUL1 has 5 isoforms with calculated MW 84-95kd and 42kd(isoform 5). This antibody can recognize isoform 1-4 .

Western Blot:A549 Cells, 1:1000-1:10000; IHC: Human ovary tumor Tissue, 1:20-1:200; IF: HepG2 Cells, 1:20-1:200; FC: HepG2 Cells, N/A

Type: Primary
Antigen: HNRNPUL1
Clonality: Polyclonal
Conjugation: Unconjugated
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Log in to see your contract pricing and availability.
  Remember me on this device


Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
150 µL 10578-1-AP 10088-392 Each (150µl) Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page


Antigen Symbol HNRNPUL1
Antigen Name Heterogeneous Nuclear Ribonucleoprotein U-like 1
Reactivity Rat, Human, Mouse
Conjugation Unconjugated
Clonality Polyclonal
Antibody Type Primary
Host Rabbit
Isotype IgG
Western Blot Yes
ImmunoChemistry Yes
ImmunoFluorescence Yes
Flow Cytometry Yes
Antigen Synonyms E1BAP5, E1B-AP5, HNRPUL1
Immunogen Recombinant Protein
Purification Antigen affinity Purification
Storage Buffer PBS with 0.1% sodium azide and 50% glycerol pH 7.3.
Shipping Temperature Store at -20°C
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty