Anti-BL21(plysS) -lysate Rabbit Polyclonal Antibody

Supplier: Proteintech
10562-1-IG-150UL
10083-620EA 459.7 USD
10083-620
Anti-BL21(plysS) -lysate Rabbit Polyclonal Antibody
Antibodies
Western Blot:Total bacteria lysates of BL21(plysS), 1:1000-1:10000

Type: Primary
Antigen: BL21(plysS) -lysate
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bacteria
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR