Anti-RANGRF Rabbit Polyclonal Antibody

Supplier: Proteintech
10552-1-AP-150UL
10093-314EA 599.76 USD
10093-314
Anti-RANGRF Rabbit Polyclonal Antibody
Antibodies
Western Blot:SGC-7901 Cells, 1:200-1:2000; IHC: Human Colon cancer Tissue, 1:20-1:200; IF: HepG2 Cells, 1:10-1:100

Type: Primary
Antigen: RANGRF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR