Anti-ERI3 Rabbit Polyclonal Antibody

10548-1-AP
10086-630EA 340.25 USD
10086-630
Anti-ERI3 Rabbit Polyclonal Antibody
Type: Primary
Antigen: ERI3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR