Anti-TAF9 Rabbit Polyclonal Antibody

Supplier: Proteintech
10544-1-AP-150UL
10095-580EA 459.7 USD
10095-580
Anti-TAF9 Rabbit Polyclonal Antibody
Antibodies
TAF9 and TAF9B are involved in transcriptional activation as well as repression of distinct but overlapping sets of genes. This antibody is a rabbit polyclonal antibody raised against the full length 172Aa isoform(AK6) of human TAF9. Adenylate kinase isoenzyme 6(AK6) has broad activity as an NMP kinase. This antibody can also recognize the 169Aa isoform, but it can not recognize the 264Aa isoform(TAF9) of this peotein.

Western Blot: HeLa Cells, 1:500-1:5000

Type: Primary
Antigen: TAF9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR