Anti-CALM3 Rabbit Polyclonal Antibody

Supplier: Proteintech
10541-1-AP
10082-106EA 360.13 USD
10082-106
Anti-CALM3 Rabbit Polyclonal Antibody
Antibodies
IHC: Human breast cancer Tissue, 1:20-1:200; Western Blot: Mouse Brain Tissue, 1:200-1:2000

Type: Primary
Antigen: CALM3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR