Anti-DPT Rabbit Polyclonal Antibody

Supplier: Proteintech
10537-1-AP-150UL
10085-394EA 459.7 USD
10085-394
Anti-DPT Rabbit Polyclonal Antibody
Antibodies
DPT (dermatopontin) is a 22 kDa extracellular matrix protein belonging to the dermatopontin family. Skin appears to be the richest source of dermatopontin, comprising about 15 mg/kg of the wet weight. Expression of dermatopontin is not limited to connective tissue, as investigations show that dermatopontin mRNA is expressed in cultured fibroblasts, muscle, heart, pancreas, and lung. Dermatopontin comprises a considerable proportion of the noncollagenous extracellular matrix proteins. It is involved in promotion of cellular adhesion, extracellular matrix assembly, wound healing, and positive modification of the growth inhibition activity of TGF-beta1.

IHC: Human pancreas cancer Tissue, 1:20-1:200; Western Blot: Mouse skin Tissue, 1:500-1:5000

Type: Primary
Antigen: DPT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR