Anti-DDR1 Rabbit Polyclonal Antibody

Supplier: Proteintech
10536-1-AP-150UL
10085-744EA 614.88 USD
10085-744
Anti-DDR1 Rabbit Polyclonal Antibody
Antibodies
IHC: Human Testis Tissue, 1:20-1:200

Type: Primary
Antigen: DDR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR