Anti-B4GALT7 Rabbit Polyclonal Antibody

Supplier: Proteintech
10535-1-AP-150UL
10083-470EA 459.7 USD
10083-470
Anti-B4GALT7 Rabbit Polyclonal Antibody
Antibodies
Western Blot: MCF7 Cells, 1:200-1:2000

Type: Primary
Antigen: B4GALT7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR