Anti-TCL1A Rabbit Polyclonal Antibody

Supplier: Proteintech
10475-1-AP-150UL
10095-734EA 459.7 USD
10095-734
Anti-TCL1A Rabbit Polyclonal Antibody
Antibodies
TCL1A, also named as TCL1 and p14 TCL1, enhances cell proliferation, stabilizes mitochondrial membrane potential and promotes cell survival. TCL1A immunodetection is an independent marker of adverse outcome that could be used in routine settings for the management of DLCL patients. TCL1A expression is correlated with shorter time to treatment in chronic lymphocytic leukemia cases and shorter lymphoma-specific survival in mantle cell lymphoma series. It is a potential therapeutic target.

Western Blot: Raji Cells, 1:200-1:1000; IHC: Human tonsillitis Tissue, 1:20-1:200

Type: Primary
Antigen: TCL1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR