Anti-GATA3 Rabbit Polyclonal Antibody

Supplier: Proteintech
10417-1-AP-150UL
10087-414EA 599.76 USD
10087-414
Anti-GATA3 Rabbit Polyclonal Antibody
Antibodies
GATA3 is a transcriptional factor that binds to the enhancer elements of all 4 T-cell antigen receptor genes. GATA3 is highly expressed in naive, freshly activated cells and Th2 lineage cells and thought to be necessary and sufficient for Th2 cytokine gene expression. GATA3 and TBET interfer with each other to regulate gene expression. This is a rabbit polyclonal antibody raised agaist part of C-terminus of human GATA3.

Western Blot: MCF7 Cells, 1:200-1:2000; IHC: Human Colon cancer Tissue, 1:20-1:200

Type: Primary
Antigen: GATA3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR