KIMBLE® Chromatography Adapter with 10-32 Thread to ¹/₄-28 Thread, DWK Life Sciences
Supplier: DWK Life Sciences
953921-6216
KT953921-6216EA 89.84 USD
KT953921-6216
KIMBLE® Chromatography Adapter with 10-32 Thread to ¹/₄-28 Thread, DWK Life Sciences
Fittings and Connectors
These adapters convert the thread style common on most HPLC systems to 1/4"-28 thread.