Would you like to log in again?
Excel Chemistry Analyzer, Stanbio Laboratory
Supplier: Stanbio Laboratory
G7700E-001
The Excel is a semi-automated clinical chemistry analyzer
Test samples are read using a flowcell module and features such as an 18-position incubator and keyboard are also included. Easy to follow instructions are displayed on the screen for each test, with additional technical support provided through our operating manual. The analyzer is equipped to handle endpoint, kinetic and EIA assays. Our open system format accommodates 120 programs and for added convenience, all Stanbio standard routine chemistries are programmed into the Excel. The analyzer completes all the necessary calculations and displays patient results in a user defined format. Printed results eliminate manual calculations and provide hard-copy patient records.
Certifications: FDA cleared.
Ordering information: Many Stanbio Reagents can be used on the Excel Chemistry Analyzer.
ORDER
Expand All / Collapse All
Description Includes Supplier No. VWR Catalog Number Unit Price Quantity
Excel Chemistry Analyzer Incubator, Keyboard, Power Adapter and Operators Manual. G7700E-001 10152-644 Each Retrieving Restricted
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:
  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address
* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
ACCESSORIES
Expand All / Collapse All
Accessories
Description Supplier No. VWR Catalog Number Unit Price Quantity
Thermal Printer Paper, 5 Rolls 3180-005 10152-646 Each Retrieving Restricted
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Q and A
Product Questions & Answers
Sort by: Most Helpful | Newest | Oldest
Items Per Page: 5  10  20 
2  3  4  Next
Can't find an answer to your question?  Ask us a question
Ask us a question:
Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question
Q&A Guidelines & Tips
If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.
When writing your question or answer, please follow these guidelines:
  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English
All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s
We reserve the right not to post your question if it contains any of the following
  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty