Anti-IgG Goat Polyclonal Antibody (DyLight® 488)

Supplier: Immunoreagents
IR2238
10150-770EA 179.53 USD
10150-770
Anti-IgG Goat Polyclonal Antibody (DyLight® 488)
Antibodies
The ImmunoReagents' DyLight® 488 conjugates provide exceptional performance for a wide array of fluorescence-base applications at a lower cost than other 488-range dyes. DyLight® 488 shows greater photostability than traditional fluorescein-based dyes and has a greater range of pH insensitivity (pH 4-9). DyLight® 488 conjugates are available for use in fluorescence microscopy, flow cytometry, Western blotting, ELISA, high-content screening and other array platforms. Excitation/Emission = 493nm / 518nm Emission Color = Green (Similar Dyes: Alexa Fluor 488, Cy2, FITC)

  • Origin: US

ImmunoReagents' antibodies are suitable for most immunoassay formats and purified, cross adsorbed and conjugated based on the final product specfications.

Products will be accompanied with a standard product CoA to include application, purity and cross reactivity profiles.

Certifications: (EC) 142/2011 approved by APHIS, Reference Number is NC-TEC-0002

Ordering information: Products can by customized based on specific assay requirements: contact your VWR representative for more information.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 488
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR