Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Goat Anti-Chicken IgY Polyclonal Antibody (DyLight® 633)

Supplier: Immunoreagents
IR1408
The ImmunoReagents' DyLight® 633 conjugates fluoresces red with physical properties comparable to other 633 dyes. DyLight® 633 is ideal for multi-color applications. DyLight® 633 is available as reactive labeling agents and as conjugates of secondary antibodies and biotin-binding proteins for use in fluorescence microscopy, flow cytometry, Western blotting, ELISA, high-content screening and other array platforms. Excitation/Emission = 638nm / 658nm Emission Color = Red (Similar Dyes: Alexa Fluor 633)

ImmunoReagents is a manufacturer of quality research and production reagents for the Life Science Industries. Product lines include a wide range of antibodies used in ELISA, immunohistochemistry (IHC), western blotting, flow cytometry (FACS), immunofluorescence and microscopy.  Antibodies produced cover a broad range of Immunoglobulins from various species cross absorbed to provide high specificity.  

ImmunoReagents' antibodies are suitable for most immunoassay formats and purified, cross adsorbed and conjugated based on the final product specfications.

Products will be accompanied with a standard product CoA to include application, purity and cross reactivity profiles.

Certifications: (EC) 142/2011 approved by APHIS, Reference Number is NC-TEC-0002

Ordering information: Products can by customized based on specific assay requirements: contact your VWR representative for more information.

Type: Secondary
Antigen: IgY
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 633
Epitope:
Host: Goat
Isotype: IgY
Reactivity: Chicken




 

ORDER

Expand All / Collapse All
Size Origin Supplier No. VWR Catalog Number Unit Price Quantity
1.0 mg US IR1408 10149-610 Each (1mg) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen Symbol IgY
Antigen Name Immunoglobulin Y
Reactivity Chicken
Conjugation DyLight® 633
Clonality Polyclonal
Antibody Type Secondary
Host Goat
Isotype IgY
Western Blot Yes
ELISA Yes
ImmunoChemistry Yes
ImmunoPrecipitation Yes
ImmunoFluorescence Yes
Flow Cytometry Yes
Concentration 1.0 mg/ml (E 1% at 280 nm = 13.0)
Format Purified IgG
Cross Adsorption No
Immunogen Chicken IgG/Y
Purification Affinity purified using solid phaseChicken IgG/Y
Form Lyophilized
Storage Buffer 10 mM Sodium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 1 % (w/v) BSA, Protease/IgG free
Storage Temperature 2-8 °C
Shipping Temperature 2-8 °C
Shelf Life 1 year from date of receipt. Prepare working dilution prior to use and then discard.
Tested Applications Fluoresence ELISA, Fluorescence-Activated Cell Sorting

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty