Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

STAT-Site® M ß-HB Meter (ß-Hydroxybutyrate System), Stanbio Laboratory

Supplier: Stanbio Laboratory
STAT-Site®
400400
The STAT-Site® M ß-HB system is intended for the quantitative determination of ß-Hydroxybutyrate in serum or plasma

It is composed of a reflectance photometer and dry-reagent test strips. This platform delivers results within 80 seconds.

Certifications: FDA cleared. CE marked.



 

ORDER

Expand All / Collapse All
Description Includes Supplier No. VWR Catalog Number Unit Price Quantity
STAT-Site® M ß-HB Meter Meter, Battery with Extra Battery Included, User's Guide, and Warranty Card 400400 10152-628 Each Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Calibration Automatic (CODE Key)
Linearity 0.0 - 2.0 mmol/L (without dilution)
Precision Low Control 6.1% CV
Sample Size 10 µL
Sample Type Serum or Plasma
Principle ß-Hydroxybutyrate dehydrogenase

ACCESSORIES

Expand All / Collapse All

Accessories
Description Supplier No. VWR Catalog Number Unit Price Quantity
STAT-Site® M ß-HB Test Strips, 4 x 10 and CODE Key 401010 10152-630 Each Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

STAT-Site® Diluent, 6 x 3 mL 202000 10152-632 Each(3ml) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

STAT-Site® Controls , 1 Low, 1 High, 2 x 3 mL 303000 10152-634 Each(3ml) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

STAT-Site® Linearity Check, 1 x 1 mL 303500 10152-636 Each (1ml) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

STAT-Site® M ß-HB Workstation 9975-001 10152-638 Each Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

STAT-Site® M ß-HB Workstation Kit with Workstation, Meter, Test Strips and CODE Key, Seropette® Pipettor (5 -50 µL) 400STATION 10152-640 Each Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

STAT-Site® M Battery Pack, Pack of 5 900905 10152-642 Each Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

STAT-Site® M ß-HB Meter Starter Pack with Meter, Test Strips, Diluent, Controls and Linearity Check 400400SP 10196-222 Each Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

STAT-Site® M ß-HB Method Comparison Set with 27 Frozen Correlation Specimens for Validation of STAT-Site M ß-HB Meter 400MC 10196-224 Each Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty