Marble, granular for producing CO₂, Supelco®
Supplier: MilliporeSigma
M-Clarity™ quality level = MQ100
1.05986.5000
1.05986.1000
1.05986.5000
EM1.05986.5000CS
742
USD
CA1.05986.5000
EM1.05986.5000
EM1.05986.1000
Marble, granular for producing CO₂, Supelco®
Marble
Formula:
CaCO₃ Storage Temperature: Ambient |
CAS Number:
471-34-1 EINECS: 207-439-9 Merck Index: 13,01658 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...