Tin(IV) chloride, Extra Pure, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Tin tetrachlorideStannic chloride

M-Clarity™ quality level = MQ200

1.07810.0500 1.07810.0500
EM1.07810.0500CS 1186 USD
CA1.07810.0500 EM1.07810.0500
Tin(IV) chloride, Extra Pure, Supelco®
Tin (IV) chloride
Formula: Cl₄Sn
MW: 260.52 g/mol
Boiling Pt: 114 °C (1013 hPa)
Melting Pt: –33 °C
Density: 2.22 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011242
CAS Number: 7646-78-8
EINECS: 231-588-9
UN: 1827
ADR: 8,II
Merck Index: 12,08929

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR