Sodium thiocyanate, pure, Supelco®

Supplier: MilliporeSigma
Warning

Synonyms: Sodium rhodanide

M-Clarity™ quality level = MQ200

1.06627.2500 1.06627.2500
EM1.06627.2500CS 1679 USD
CA1.06627.2500 EM1.06627.2500
Sodium thiocyanate, pure, Supelco®
Sodium thiocyanate
Formula: NaSCN
MW: 81.07 g/mol
Melting Pt: 287 °C
Density: 1.735 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011123
CAS Number: 540-72-7
EINECS: 208-754-4
Merck Index: 12,09463

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR