Sodium cyanide, pure, Supelco®
Supplier: MilliporeSigma
|
Danger
|
M-Clarity™ quality level = MQ200
1.06437.1000
1.06437.9050
1.06437.1000
1.06437.9050
EM1.06437.1000CS
1862.19
USD
EM1.06437.1000
CA1.06437.1000
EM1.06437.9050
CA1.06437.9050
Sodium cyanide, pure, Supelco®
Sodium cyanide
Formula:
CNNa MW: 49.01 g/mol Boiling Pt: 1496 °C (1013 hPa) Melting Pt: 563.9 °C Density: 1.595 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003523 CAS Number: 143-33-9 EINECS: 205-599-4 UN: 1689 ADR: 6.1,I Merck Index: 12,08750 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...