Potassium cyanide, Supelco®
Supplier: MilliporeSigma
|
Danger
|
M-Clarity™ quality level = MQ200
1.04965.1000
1.04965.1000
EM1.04965.1000CS
1084
USD
EM1.04965.1000
CA1.04965.1000
Potassium cyanide, Supelco®
Potassium cyanide
Formula:
KCN MW: 65.12 g/mol Boiling Pt: 1625 °C (1013 hPa) Melting Pt: 634 °C Density: 1.55 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011397 CAS Number: 151-50-8 EINECS: 205-792-3 UN: 1680 ADR: 6.1,I Merck Index: 12,07789 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...