Isoamyl acetate, Supelco®

Supplier: MilliporeSigma
Warning

Synonyms: Isopentyl acetateAcetic acid isoamyl ester

M-Clarity™ quality level = MQ200

1.01231.1000 1.01231.6025 1.01231.6025
EM1.01231.6025EA 1793 USD
CA1.01231.6025 EM1.01231.6025 EM1.01231.1000
Isoamyl acetate, Supelco®
Isoamyl acetate
Formula: C₇H₁₄O₂
MW: 130.19 g/mol
Boiling Pt: 142 °C (1013 hPa)
Melting Pt: –78 °C
Density: 0.87 g/cm³ (20 °C)
Flash Pt: 25 °C
Storage Temperature: Ambient
MDL Number: MFCD00008946
CAS Number: 123-92-2
EINECS: 204-662-3
UN: 1104
ADR: 3,III
Merck Index: 12,05125

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR