Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
  • Supplier
  • Suppliers found in search results
    Sort by:

  • Search Within Results

You Searched for: "rnai"

https://us.vwr.com/stibo/smallweb/std.lang.all/96/98/18989698.jpg

RNA interference is a natural process that cells use to silence unwanted or harmful genes, particularly if the cell is threatened or damaged by invading viruses. Controls and inhibitors are available for siRNAs and miRNAs. These controls are specially designed to be used as positive, negative, or transfection influencers for validation of gene-silencing experiments. Certain control siRNAs are fluorescently labeled for the best monitoring transfection efficiency.

SearchPresentationType-VERTICAL
 
 
SearchResultCount:"7"
Sort by:
 
 
 
 

Supplier:  Genlantis
Description:   The Dicer siRNA Generation Kit allows easy generation of a large number of small interfering RNAs (siRNAs) from full-length target genes. This kit mim...
Supplier:  Genlantis
Description:   Turbo Dicer™ siRNA Generation kit uses an ultra-active form of human recombinant dicer enzyme that can cleave more than 95% of dsRNA template into 22 ...
Supplier:  Bioneer
Description:   AccuTarget™ Control siRNAs are a wide range of positive control siRNAs, negative control siRNAs and control siRNA sets. Control siRNA can be fluoresce...
Supplier:  Bioneer
Description:   AccuTarget™ Human siRNA Library contains 25 pathway specific siRNA for researchers studying cellular processes, cancer, disease etc. These are availab...
Supplier:  Bioneer
Description:   AccuTarget™ Human siRNA subset libraries are functional/family subsets of the Kinase or phosphatase libraries. These are available at 0.1, 0.25, 0.5 a...
Supplier:  Bioneer
Description:   1 nmole, bio-rp purifiedl. validated siRNAs are proven to show 80[perCent] knockdown efficiency.
Supplier:  New England Biolabs (NEB)
Description:   The p19 siRNA Binding Protein (19 kDa) from the Carnation Italian Ringspot Virus (CIRV) plant binds siRNAs with nanomolar affinity. The dimeric protei...
Small Business Enterprise
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

???store.product.restricted???
This product has been blocked by your organization.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at 1-800-932-5000.
1 - 7  of 7