Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

DRIERITE® Gas Purifiers, Hammond

Supplier: HAMMOND W.A. DRIERITE
27068
This DRIERITE Gas Purifier is specially designed for gas chromatography and other applications requiring dry, pure gas.

  • Ideal for use in gas chromatography to prevent ghost peaks and to improve baselines

It will dry, purify, and filter gases used for chromatography and spectrometry

This unit is available filled with 500 cc of Indicating DRIERITE for applications that require only drying and filtering. The gas purifier should be used for carrier gases and for flame lonization detector air and hydrogen gases. The gas purifier may also be used to remove moisture and impurities from gases when purging spectrophotometer compartments,etc.

Indicating DRIERITE dries gases to a dryness of 0.005 mg/l. Color changes from blue to pink upon exhaustion. 5A Molecular Sieves remove impurities that have an effective molecular diameter of less than 5 angstroms.

The purifier is filled with Indicating DRIERITE and 5A Molecular Sieves for the removal of moisture, impurities, and particulates from gas lines. It is easy to attach using the compression type tube fittings common to most laboratory instruments.

Ordering information: Plastic coated mounting clips may be purchased separately; contact your VWR representative for more information. For use in gas chromatography to prevent ghost peaks and to improve baselines.



 

ORDER

Expand All / Collapse All
Description Capacity Supplier No. VWR Catalog Number Unit Price Quantity
Model L68GP Gas Purifier 25 g (Water Capacity) 27068 22891-090 Case of 6 Retrieving Restricted
Each Retrieving Restricted
View MSDS SDS
restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Max. Operating Pressure (bar) 6.9
Ø×L (mm) 2 5/8 x 11 3/8"
Recommended Flow Rate up to 300 L/hour

ACCESSORIES

Expand All / Collapse All

Accessories
Description Supplier No. VWR Catalog Number Unit Price Quantity
Gas Purifier Refill Kit RF68 27069 22891-092 Case of 6 Retrieving Restricted
Each Retrieving Restricted
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty