Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Whatman Grade 0860 ½ Qualitative Filter Papers, GE Healthcare

Supplier: GE Healthcare - Whatman
10334547 10334553 10334551
Savings in time required to quadrant-fold circles to fit conical filter funnels in repetitive or multiple analyses.

  • Increased total loading capacity as more filter area is available

Grade 0860 ½ filter papers are folded, comparable to Grade 0858 ½ but with a smooth surface, slightly thinner and faster. Decreased overall filtration time because of the extra surface area exposed; the normal slow down of filtration speed due to the loading of particulate is postponed.

Maintained flow rate due to the reduction in filter paper contact with funnel side and the self-supporting shape of the filter itself.

The prepleating does not significantly affect any of the technical data, and the same figures may be used for the flat circles.

Time-saving Whatman qualitative grades are offered in this convenient form, which gives major advantages over flat circles.

Comparable to Grade 0858 but with a smooth surface, slightly thinner and faster. Also available flat stock form as Grade 0860.




 

ORDER

Expand All / Collapse All
Description Diameter Filtration Speed Supplier No. VWR Catalog Number Unit Price Quantity
Grade 0860 ½ Qualitative Filter Paper Folded (Prepleated), 185 mm 185 mm 60 s (Herzberg) 10334547 10034-756 Pack of 100 Retrieving
Product available on GSA Advantage®
Grade 0860 ½ Qualitative Filter Paper Folded (Prepleated), 240 mm 240 mm 60 s (Herzberg) 10334551 10034-758 Pack of 100 Retrieving
Product available on GSA Advantage®
Grade 0860 ½ Qualitative Filter Paper Folded (Prepleated), 320 mm 320 mm 60 s (Herzberg) 10334553 89233-664 Pack of 100 Retrieving
Product available on GSA Advantage®
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Weight 75 g/m2
Thickness 170 µm
Particle Retention 12 µm ( in Liquid Max.)
Filtration Speed Approx. 60 s (Herzberg)
Basis Weight 75 g/m²

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty