Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

VWR® Standard Stirrers, 230V (Export Only)

Supplier: VWR International
VWR®
984VW4ASTEUS 984VW0CSTEUS 984VW7CSTEUS 984VW4CSTEUS 984VW0ASTEUS 984VW7ASTEUS
Standard stirrers are designed for general-purpose laboratory use, and these analog units are microprocessor controlled and are equipped with easy-to-use, dial-adjustment controls.

  • Enhanced electronics
  • Cool touch, chemical-resistant housing
  • Best-in-Class Five-Year Warranty

Enhanced electronics regulate stirring functions

Stirrers are available with ceramic or aluminum top plates. Ceramic top plates feature an easy-to-clean, chemical-resistant, reflective white surface (units offer ±3% temperature stability). Aluminum top plates will not crack or chip (units offer ±2% temperature stability). The unit's housing is constructed of a polymer that is chemical-resistant.

A ramping feature gradually increases speed to prevent splashing, improve magnetic coupling, and provide excellent low end control. Speed is precisely controlled, with consistent stirring at all speeds powered by a dependable, continuous-duty motor. A low-profile design requires minimal bench space and permits use within a fume hood. The spill-resistant housing channels fluids away from internal components in the event of a spill.

Certifications: RoHS certified. TUV listed.

Ordering information: Units are supplied with a 234 cm (92") Euro type plug. A 3.8 cm (1.5") PTFE-coated stir bar is also included. The optional support rod and clamp kit includes a 45.7 cm (18") long stainless steel support rod, a thermometer/temperature probe extension clamp, a three-prong dual-adjust swivel clamp, and a hook connector.



 

ORDER

Expand All / Collapse All
Overall Dimensions Top Plate Dimensions Maximum Capacity Top Plate Material Speed Range Shipping Weight VWR Catalog Number Unit Price Quantity
27.4 x 16.8 x 10.9 cm (10.8 x 6.6 x 4.3") 10.2 x 10.2 cm (4 x 4") 600 mL Ceramic 60–1600 rpm 2.8 kg (6.2 lbs.) 97042-588 Each Retrieving
Product available on GSA Advantage® CE Compliant
37.6 x 25 x 10.9 cm (14.8 x 9.9 x 4.3") 17.8 x 17.8 cm (7 x 7") 2500 mL Ceramic 60–1600 rpm 4.8 kg (10.5 lbs.) 97042-628 Each Retrieving
Product available on GSA Advantage® CE Compliant
37.6 x 25 x 10.9 cm (14.8 x 9.9 x 4.3") 17.8 x 17.8 cm (7 x 7") 2500 mL Aluminum 60–1600 rpm 4.8 kg (10.5 lbs.) 97042-632 Each Retrieving
Product available on GSA Advantage® CE Compliant
45.5 x 33 x 10.9 cm (17.9 x 13 x 4.3") 25.4 x 25.4 cm (10 x 10") 6000 mL Ceramic 60–1600 rpm 7.2 kg (15.9 lbs.) 97042-668 Each Retrieving
Product available on GSA Advantage® CE Compliant
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Environmentally Preferable Attribute
Speed stability +/- 2%
Electrical 230V, 0.2A, 50W
Environmental Certification

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty