VWR® Professional Stirrers, 230 V (Export Only)
Supplier: VWR, part of Avantor
984VW7AHSEUP 984VW0AHSEUP-C 984VW7CHSEUP 984VW7AHSEUP-C 984VW7CHSEUP-C 984VW0CHSEUP-C 984VW0CHSEUP
97042-756EA 2137.98 USD
97042-756 10027-066 10027-054 97042-716 97042-724 10027-058 10027-062
VWR® Professional Stirrers, 230 V (Export Only)
Designed for applications requiring exceptional accuracy, stability, and repeatability, these hotplate stirrers are equipped with an exclusive safety system that protects both the operator and sample.
  • Excellent temperature uniformity
  • LED displays for temperature, speed, and time
  • Cool touch, chemical-resistant housing
  • Includes RTD temperature probe kit
  • Best-in-Class Five-Year Warranty
  • Available with a NIST Traceable, ISO/IEC 17025 Accredited Calibration Certificate from Troemner
  • Industry Leading Safety Features
Easy-to-read touch pad controls are located on the front of the unit, with independent LED displays for temperature, speed, and time
The low-profile design occupies minimal bench space, and fits into fume hoods. The rear housing of the unit features an off-centered, built-in support rod holder with locking knob that accepts the probe kit supplied with each unit. Hotplate stirrers are available with ceramic or aluminum top plate material. White reflective ceramic tops are chemical-resistant and easy to clean. Durable aluminum tops will not crack or chip, and offer a more even heating surface.
Hotplate stirrers are equipped with a variety of innovative safety features designed to facilitate efficient operation and workplace safety. Unit housing is constructed of a heat- and chemical-resistant polymer that remains cool to the touch throughout use. The spill-resistant housing channels fluids away from internal components in the event of a spill. A maximum temperature limiting function ensures the plate temperature will not exceed preset limits, allowing for control of sensitive flash points. A hot symbol warning light is illuminated when the heat is turned on, and remains lit until the top plate is sufficiently cooled. In timed mode, an alarm will sound when time reaches zero, or when the unit reaches set point temperature. Additionally, if the probe disengages from the sample, or if the stirrer motor stops or fails, the heater will automatically shut off. Units are equipped with redundant safety controls, which will shut down unit in case of heater malfunction
A timer displays elapsed time or a user-defined limit, and will shut off the unit when time reaches zero. The last used settings will be displayed, even after the power has been shut off. Enhanced electronics regulate the heating and stirring, and will bring samples to the desired temperature quickly and efficiently. An RTD ensures precise temperature control. A powerful continuous duty motor delivers dependable and reliable stirring at all speeds.
Certifications: RoHS certified. TUV listed. Calibrated Hotplate-Stirrers include a Troemner ISO/IEC Accredited Calibration Certificate traceable to NIST. The ISO/IEC 17025 Accredited Calibration Certificate includes multiple data points within the temperature and speed ranges and time function with the associated uncertainties. Troemner is accredited by NVLAP under Laboratory Code 105013-0 to perform ISO/IEC 17025 calibrations.
Ordering information: Units include a 234 cm (92") Euro type plug. Unit is also supplied with a 3.8 cm (1.5") PTFE-coated stir bar. Units are also supplied with a probe kit which includes a 20.3 cm (8") stainless steel RTD temperature probe, 45.7 cm (18") stainless steel support rod, thermometer/temperature probe extension clamp, and hook connector. Hotplate Stirrers with NIST traceable calibration certificate provided by an ISO/IEC 17025 accredited laboratory. Multiple data points within the temperature range and time function are reported on the certificate with the associated uncertainties.