VWR® Advanced Slow Speed Stirrers, 230 V (Export Only)
Supplier: VWR, part of Avantor
986937
12621-078EA 1430.03 USD
12621-078
VWR® Advanced Slow Speed Stirrers, 230 V (Export Only)
Designed for growth of suspension and micro-carrier cell cultures, slow speed stirrers work with cell culture flasks and other vessels designed for slow magnetic stirring.
  • Ideal for Cell Culture Applications
  • Slow Start/Stop Feature
  • Efficient Drive Motor Eliminates Heat Transfer to Sample
  • Best-in-Class Five-Year Warranty
  • Available with a NIST Traceable, ISO/IEC 17025 Accredited Calibration Certificate from Troemner
Ideal for use in cold rooms, incubators, or CO2 incubators (-10 to 40°C), 20-80% relative humidity, non-condensing environment. The low-profile design takes up minimal space and easily fits into fume hoods and most incubators. Cast aluminum base offers durability and added stability. Unit features a chemical resistant, reflective white top plate surface that is easy to clean. Microprocessor control precisely regulates speed through the entire range. Stirring function, with continuous duty motor and magnet, maintains set speed even under changing load or viscosity.
Certifications: RoHS certified. Calibrated Stirrers include a Troemner ISO/IEC Accredited Calibration Certificate traceable to NIST. The ISO/IEC 17025 Accredited Calibration Certificate includes multiple data points within the speed range and time function with the associated uncertainties. Troemner is accredited by NVLAP under Laboratory Code 105013-0 to perform ISO/IEC 17025 calibrations.
Ordering information: Unit includes a 234 cm (92") Euro type plug.