Ergocalciferol (vitamin D2, calciferol) 97.0-103.0%

Danger

Synonyms: (S,Z)-3-((E)-2-((1R,3aS,7aR)-1-((2R,5R,E)-5,6-Dimethylhept-3-en-2-yl)-7a-methylhexahydro-1H-inden-4(2H)-ylidene)ethylidene)-4-methylenecyclohexanol

J15562-06 J15562-14
101172-472EA 145.25 USD
101172-472 101174-360
Ergocalciferol (vitamin D2, calciferol) 97.0-103.0%
Ergocalciferol (Vitamin D2, Calciferol)
Formula: C₂₈H₄₄O
MW: 396.66 g/mol
Melting Pt: 118…119 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00166988
CAS Number: 50-14-6
EINECS: 200-014-9
UN: 2811
ADR: "6.1",III
Merck Index: 14,10018

Order Now

Specification Test Results

Assay (unspecified) 97.0 - 103.0% (as is)
Assay (unspecified - Reducing Substances) Passes Test
Identification (A, B, C, D) Passes Test
Optical Rotation (c= 1.5 in EtOH) +103 to 106°

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR