UltraSonic Cutter, Electron Microscopy Sciences
55180-380
101412-018EA 19947.34 USD
101412-018
UltraSonic Cutter, Electron Microscopy Sciences
The UltraCut™ 380 Ultrasonic Cutter is a specialized cutting instrument which utilizes abrasive powders and high speed, vibratory motion to cut discs, holes, cylinder, squares and others
The high speed motion is developed using an ultrasonic transducer oscillating at 26kH. Boron carbide slurry is applied to the sample and the tool is lowered onto the specimen. The particles under the vibrating tool impact the sample and erode the material in the shape of the tool.