2-Oxoglutaric acid free acid ≥98%

Danger

Synonyms: 2-Ketoglutaric acid2-Oxopentanedioic acidA-Ketoglutaric acidα-Ketoglutaric acid

J17950-36
101169-934EA 164.77 USD
101169-934
2-Oxoglutaric acid free acid ≥98%
2-Oxoglutaric acid
Formula: HOOCCH₂CH₂COCOOH
MW: 146.1 g/mol
Boiling Pt: 323 °C (1013 hPa)
Melting Pt: 113…117 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004165
CAS Number: 328-50-7
EINECS: 206-330-3
Merck Index: 14,05303

Order Now

Specification Test Results

Assay (unspecified) ≥98.0% (as is)
Identification (FTIR) Conforms
Loss on Drying ≤1.0%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR