KIMBLE® Straight Flow Control Inlet Adapter with PTFE Stopcock, DWK Life Sciences
Supplier: DWK Life Sciences
186055-2440
KT186055-2440EA 189.84 USD
KT186055-2440
KIMBLE® Straight Flow Control Inlet Adapter with PTFE Stopcock, DWK Life Sciences
Adapters Adapters, Joints with Tube Connectors
Use this adapter for regulating the input of gases or liquids into reaction systems.
  • Stopcock with PTFE plug
  • Inner [ST] joint
  • With hose connection
Adapter features an inner Standard Taper joint
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
The hose connection accepts ⅜” ID flexible tubing. Stopcock has a PTFE plug.