You Searched For: Specimen Mounting Supplies
Choose from a full array of products designed for insect specimen mounting, including spreading boards, mounting pins, mounting paper, pinning blocks, and holding fluids for preserving specimens without the use of formaldehyde. Complete mounting kits are also available for mounting and displaying insects. Labels and pins can be purchased independently to supplement larger packages, and pins have extremely sharp points and small heads to ensure precise pinning without tearing mounting specimens.
Choose from a full array of products designed for insect specimen mounting, including spreading boards, mounting pins, mounting paper, pinning blocks, and holding fluids for preserving specimens without the use of formaldehyde. Complete mounting kits are also available for mounting and displaying insects. Labels and pins can be purchased independently to supplement larger packages, and pins have extremely sharp points and small heads to ensure precise pinning without tearing mounting specimens.
169  results were found
SearchResultCount:"169"
Sort Results
List View Easy View
Rate These Search Results
Catalog Number: (10066-630)
Supplier: VWR International
Description: The PolyScience Histology Freeze Plate is a fast-freeze cold plate that allows histotechnicians to observe tissue freezing and keep specimen edges fla...
Catalog Number: (470149-222)
Supplier: VWR International
Description: Insect Pinning Block
Supplier: Epredia
Description: Get the most out of your Thermo Scientific™ microtome with this Thermo Scientific™ Specimen Clamp and Accessories.
Supplier: Epredia
Description: Employ chrome–plated disks precisely machined to accurate dimensions with Thermo Scientific™ Shandon™ Specimen Object Disks.
Supplier: PERKINELMER U.S. LLC
Description: Rectangular windows make the demountable cell becomes a semi-demountable cell. It can be used either as a demountable cell or as a sealed cell for hig...
Catalog Number: (10139-110)
Supplier: PERKINELMER U.S. LLC
Description: Silver Bromide circular demountable cell windows are minimally degraded by water making them useful for applications in the deep infrared.  
Supplier: PERKINELMER U.S. LLC
Description: Circular windows are used to construct a demountable cell where the sample is placed on one window and the second window is pressed lightly on top of ...
Catalog Number: (10136-238)
Supplier: PERKINELMER U.S. LLC
Description: Zinc Selenide (Irtran-4™) is similar to Cleartran except that it is somewhat softer and more easily scratched.
Supplier: United Scientific Supplies
Description: These steel pins are coated with a corrosion-resistant finish and have a yellow epoxy head.
Catalog Number: (101413-632)
Supplier: Electron Microscopy Sciences
Description: Aluminum slotted head with 3.1 mm diameter pin, fits most SEMs.
Minority or Woman-Owned Business Enterprise
Supplier: Electron Microscopy Sciences
Description: Boxes in five configurations are available for storing SEM pin stub mounts and cylinder mounts.
Minority or Woman-Owned Business Enterprise
Supplier: Electron Microscopy Sciences
Description: EMS Mica Sheets offer a clean surface for EM applications, carbon filming and particle spraying, as well as for AFM applications. The highest quality ...
Supplier: Electron Microscopy Sciences
Description: These stainless steel chucks are designed to be used cold for rapid freezing, and to maximize the gripping power required to hold the embedded tissue ...
Supplier: Electron Microscopy Sciences
Description: Slotted head pin stub mounts with tappered pin ends are for use with Cambridge, Phillips, Camscan, B and L, Etec, Zeiss.
Minority or Woman-Owned Business Enterprise
Supplier: Electron Microscopy Sciences
Description: Specially designed for easy, quick and clean mounting of any type of thin specimen on specimen stubs.
Supplier: SPRINGSIDE SCIENTIFIC, LLC.
Description: Suitable for Leica (except for new model CM1950), TBS, Tanner, older Avantik and other common cryostats
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at 1-800-932-5000.
1 - 16 of 169
no targeter for Bottom