Corning® Gosselin™ S-Blender-1A, Corning
Supplier: Corning
S-BLENDER-1A
10152-748CS 7508.78 USD
10152-748
Corning® Gosselin™ S-Blender-1A, Corning
The Gosselin™ S-Blender-1A is a high performance blender designed for microbiological sample preparation
The blender, which is suitable for a range of sample dilutions between 50mL and 400mL, ensures quick and accurate blending of any food, agricultural, chemical, or cosmetic sample.