Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Azure™ cSeries Advanced Imaging Systems, Azure Biosystems

Supplier: Azure Biosystems
AC3001 AC1026 AC6001 AC4001 AC5001 AC1084
The Azure Biosystems cSeries offers four unique imaging systems. All systems offer a wide range of applications with multiple light sources and filters.

  • For a wide range of applications
  • Multiple light sources and filters
  • Qualification packages and services available

The c200 gel imaging workstation can image multiple types of gels

The Darkroom Replacer, Model c300, features fast chemiluminescent detection with no film or darkroom needed. The c300 can be upgraded to any of the ascending cSeries models (c400, c500 or c600) with a simple field upgrade.

The Visible Fluorescent Western Imaging System, Model c400, features a three-channel RGB excitation for Cy5/Cy3/CY2 Western applications. It also features fast chemiluminescent detection with no film or darkroom needed. The c400 can be upgraded to an IR imaging system (Model c600) with a simple field upgrade.

The Infrared Imaging System, Model c500, features two-channel IR direct detection with laser diodes at 660 nm and 785 nm. Narrow-band excitation enables NIR applications with better sensitivity than LEDs or white light sources. The c500 can image multiple types of gels and blots. It also features fast chemiluminescent detection with no film or darkroom needed. The c500 can be upgraded to the c600 with a simple field upgrade.

The included UV and white light transillumination allows fluorescent and white-light imaging of gels, including nucleotide gels stained with ethidium bromide and protein gels stained with Coomassie blue or silver stain. It provides white and blue epi illumination for other types of gels and blots such as SYBR safe. It can be upgraded to any of the ascending models (c300, c400, c500 or c600). The system comes with AzureSpot analysis software.

Certifications: UL, CSA and CE compliant.

Accessories information: The Installation, Operational, and Performance Qualification (IQOQPQ) package includes the protocols, checklist, targets, and certificates necessary to satisfy your validation requirements. The simple to follow instructions and durable Checklist & Logbook will assist you in performing the necessary tests, and maintaining accurate records.




 

ORDER

Expand All / Collapse All

cSeries Imaging Systems
Description Model Wavelength Supplier No. VWR Catalog Number Unit Price Quantity
The Darkroom Replacer Imaging System c300 302, 365 460 nm AC3001 10147-220 Each Retrieving

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Product available on GSA Advantage®
The Visible Fluorescent Western Imaging System c400 302, 365 460, 470 526, 628 nm AC4001 10147-218 Each Retrieving
Product available on GSA Advantage®
The Infrared Imaging System c500 302, 365 460, 660, 785 nm AC5001 10147-216 Each Retrieving

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Product available on GSA Advantage®
The Ultimate Western Blot Imaging System c600 302, 365 460, 470, 526, 628, 660, 785 nm AC6001 10147-214 Each Retrieving

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Product available on GSA Advantage®

Qualifications
Description Supplier No. VWR Catalog Number Unit Price Quantity
IQOQPQ procedure for c300-c600 AC1026 75794-846 Each Retrieving

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Product available on GSA Advantage®

Warranties and Services for c300 Imaging Systems
Description Supplier No. VWR Catalog Number Unit Price Quantity
1-Year Extended Warranty for c300 AC1084 10147-266 Each Retrieving
Product available on GSA Advantage®
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

W×D×H 38 x 36 x 55 cm
Field of View 20×15 cm

ACCESSORIES

Expand All / Collapse All

Accessories
Description Supplier No. VWR Catalog Number Unit Price Quantity
Chemi Tray AC1017 10147-254 Each Retrieving
Gel Carrying Tray AC1023 10001-876 Each Retrieving
Gel Cutting Screen AC1024 10001-818 Each Retrieving
IR Package Upgrade Kit: Laser Excitation Modules for Laser Diodes (660 nm and 785 nm), Emmission Filters (720 nm and 815 nm) AC1033 10147-274 Each Retrieving

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Orange Tray AC1016 10147-252 Each Retrieving
RGB Package AC1032 10147-272 Each Retrieving

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Single License AC1061 10147-224 Each Retrieving

Warranties and Services for c400 Imaging Systems
Description Supplier No. VWR Catalog Number Unit Price Quantity
Preventative Maintenance for c300, c400, c500, and c600 AC1112 75996-612 Each Retrieving

Warranties and Services for c500 Imaging Systems
Description Supplier No. VWR Catalog Number Unit Price Quantity
Preventative Maintenance for c300, c400, c500, and c600 AC1112 75996-612 Each Retrieving

Warranties and Services for c600 Imaging Systems
Description Supplier No. VWR Catalog Number Unit Price Quantity
Preventative Maintenance for c300, c400, c500, and c600 AC1112 75996-612 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty